missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[N-[alpha-[2-(Piperidinoacetamido)phenyl]benzylidene]glycinato]nickel 93.0+%, TCI America™
Supplier: TCI America P17371G
Specifications
| [N-[alpha-[2-(Piperidinoacetamido)phenyl]benzylidene]glycinato]nickel | |
| C22H23N3NiO3 | |
| WUBYDZLSGCSGEK-UHFFFAOYSA-L | |
| nickel(2+);2-[[[2-[(1-oxido-2-piperidin-1-ylethylidene)amino]phenyl]-phenylmethylidene]amino]acetate | |
| 25231255 | |
| ≥93.0% (T) |
| 847654-17-5 | |
| 1 g | |
| C1CCN(CC1)CC(=NC2=CC=CC=C2C(=NCC(=O)[O-])C3=CC=CC=C3)[O-].[Ni+2] | |
| 436.137 | |
| 436.14 | |
| Crystalline Powder |
Chemical Identifiers
| 847654-17-5 | |
| 436.137 | |
| 25231255 | |
| C1CCN(CC1)CC(=NC2=CC=CC=C2C(=NCC(=O)[O-])C3=CC=CC=C3)[O-].[Ni+2] |
| C22H23N3NiO3 | |
| WUBYDZLSGCSGEK-UHFFFAOYSA-L | |
| nickel(2+);2-[[[2-[(1-oxido-2-piperidin-1-ylethylidene)amino]phenyl]-phenylmethylidene]amino]acetate |
Safety and Handling
TSCA : No