missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[N-[alpha-[2-(Piperidinoacetamido)phenyl]benzylidene]glycinato]nickel 93.0+%, TCI America™
$68.97 - $380.83
Chemical Identifiers
| CAS | 847654-17-5 |
|---|---|
| Molecular Formula | C22H23N3NiO3 |
| Molecular Weight (g/mol) | 436.137 |
| InChI Key | WUBYDZLSGCSGEK-UHFFFAOYSA-L |
| PubChem CID | 25231255 |
| IUPAC Name | nickel(2+);2-[[[2-[(1-oxido-2-piperidin-1-ylethylidene)amino]phenyl]-phenylmethylidene]amino]acetate |
| SMILES | C1CCN(CC1)CC(=NC2=CC=CC=C2C(=NCC(=O)[O-])C3=CC=CC=C3)[O-].[Ni+2] |
Chemical Identifiers
| 847654-17-5 | |
| 436.137 | |
| 25231255 | |
| C1CCN(CC1)CC(=NC2=CC=CC=C2C(=NCC(=O)[O-])C3=CC=CC=C3)[O-].[Ni+2] |
| C22H23N3NiO3 | |
| WUBYDZLSGCSGEK-UHFFFAOYSA-L | |
| nickel(2+);2-[[[2-[(1-oxido-2-piperidin-1-ylethylidene)amino]phenyl]-phenylmethylidene]amino]acetate |
Specifications
| 847654-17-5 | |
| 100 mg | |
| C1CCN(CC1)CC(=NC2=CC=CC=C2C(=NCC(=O)[O-])C3=CC=CC=C3)[O-].[Ni+2] | |
| 436.137 | |
| 436.14 | |
| Crystalline Powder |
| C22H23N3NiO3 | |
| WUBYDZLSGCSGEK-UHFFFAOYSA-L | |
| nickel(2+);2-[[[2-[(1-oxido-2-piperidin-1-ylethylidene)amino]phenyl]-phenylmethylidene]amino]acetate | |
| 25231255 | |
| ≥93.0% (T) | |
| [N-[alpha-[2-(Piperidinoacetamido)phenyl]benzylidene]glycinato]nickel |
Safety and Handling
TSCA : No