missing translation for 'onlineSavingsMsg'
Learn More

N-Allylthiourea, 98%

CAS: 109-57-9 | C4H8N2S | 116.182 g/mol

Supplier:  Thermo Scientific Chemicals L0337714

 View more versions of this product

Catalog No. AAL0337714


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
N-Allylthiourea is a nitrification inhibitor used in the study on the transformation of diclofenac, naproxen and bisoprolol under aerobic and anaerobic conditions. It is also used in medicine to minimize scar tissue in order to fight against a type of dermatitis. Further, it inhibits the growth of transplanted tumors in mice. It acts as a chelating agent. In addition, it is used in cosmetics, preservative and in organic synthesis.

Solubility
Soluble in water and alcohol. Slightly soluble in diethyl ether. Insoluble in benzene.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

N-Allylthiourea
70°C to 78°C
Unpleasant
MFCD00004940
UN2811
14,9362
Soluble in water and alcohol. Slightly soluble in diethyl ether. Insoluble in benzene.
C=CCNC(=S)N
116.182
CHEBI:74079
98%
109-57-9
1.22
C4H8N2S
25 g
1071470
allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin
HTKFORQRBXIQHD-UHFFFAOYSA-N
prop-2-enylthiourea
1549517
116.19

Chemical Identifiers

109-57-9
116.182
HTKFORQRBXIQHD-UHFFFAOYSA-N
1549517
prop-2-enylthiourea
C4H8N2S
MFCD00004940
allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin
CHEBI:74079
C=CCNC(=S)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 203-683-5

RTECSNumber : YR8050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only