missing translation for 'onlineSavingsMsg'
Learn More

N-Allylthiourea, 98%

Catalog No. AAL0337722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0337722 100 g
AAL0337714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0337722 Supplier Thermo Scientific Chemicals Supplier No. L0337722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 109-57-9 | C4H8N2S | 116.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
N-Allylthiourea is a nitrification inhibitor used in the study on the transformation of diclofenac, naproxen and bisoprolol under aerobic and anaerobic conditions. It is also used in medicine to minimize scar tissue in order to fight against a type of dermatitis. Further, it inhibits the growth of transplanted tumors in mice. It acts as a chelating agent. In addition, it is used in cosmetics, preservative and in organic synthesis.

Solubility
Soluble in water and alcohol. Slightly soluble in diethyl ether. Insoluble in benzene.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 109-57-9
Molecular Formula C4H8N2S
Molecular Weight (g/mol) 116.182
MDL Number MFCD00004940
InChI Key HTKFORQRBXIQHD-UHFFFAOYSA-N
Synonym allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin
PubChem CID 1549517
ChEBI CHEBI:74079
IUPAC Name prop-2-enylthiourea
SMILES C=CCNC(=S)N

Specifications

Melting Point 70°C to 78°C
Density 1.22
Odor Unpleasant
Quantity 100 g
UN Number UN2811
Beilstein 1071470
Merck Index 14,9362
Solubility Information Soluble in water and alcohol. Slightly soluble in diethyl ether. Insoluble in benzene.
Formula Weight 116.19
Percent Purity 98%
Chemical Name or Material N-Allylthiourea
Show More Show Less
Hazard Category H301
Hazard Statement GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 203-683-5
RTECSNumber YR8050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only