missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Allyloxyphthalimide 98.0+%, TCI America™
$65.43 - $348.57
Chemical Identifiers
| CAS | 39020-79-6 |
|---|---|
| Molecular Formula | C11H9NO3 |
| Molecular Weight (g/mol) | 203.20 |
| MDL Number | MFCD00173307 |
| InChI Key | XVKREICBUWCANY-UHFFFAOYSA-N |
| PubChem CID | 2765097 |
| IUPAC Name | 2-(prop-2-en-1-yloxy)-2,3-dihydro-1H-isoindole-1,3-dione |
| SMILES | C=CCON1C(=O)C2=CC=CC=C2C1=O |
Chemical Identifiers
| 39020-79-6 | |
| 203.20 | |
| XVKREICBUWCANY-UHFFFAOYSA-N | |
| 2-(prop-2-en-1-yloxy)-2,3-dihydro-1H-isoindole-1,3-dione |
| C11H9NO3 | |
| MFCD00173307 | |
| 2765097 | |
| C=CCON1C(=O)C2=CC=CC=C2C1=O |
Specifications
| 39020-79-6 | |
| C11H9NO3 | |
| 5 g | |
| C=CCON1C(=O)C2=CC=CC=C2C1=O | |
| 203.20 | |
| 203.20 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00173307 | |
| XVKREICBUWCANY-UHFFFAOYSA-N | |
| 2-(prop-2-en-1-yloxy)-2,3-dihydro-1H-isoindole-1,3-dione | |
| 2765097 | |
| ≥98.0% (GC) | |
| N-Allyloxyphthalimide |
Safety and Handling
TSCA : No