accessibility menu, dialog, popup

UserName

CAS RN 39020-79-6

H2CONOO

CAS RN 39020-79-6

IUPAC Name: 2-(prop-2-en-1-yloxy)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(prop-2-en-1-yloxy)isoindole-1,3-dione
Molecular Weight (g/mol): 203.2
Molecular Formula: C11H9NO3
InChi Key: XVKREICBUWCANY-UHFFFAOYSA-N
SMILES: C=CCON1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results