missing translation for 'onlineSavingsMsg'
Learn More

N-Allyloxyphthalimide 98.0+%, TCI America™

Catalog No. A14475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14475G 5 g
A144725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14475G Supplier TCI America Supplier No. A14475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 39020-79-6
Molecular Formula C11H9NO3
Molecular Weight (g/mol) 203.20
MDL Number MFCD00173307
InChI Key XVKREICBUWCANY-UHFFFAOYSA-N
PubChem CID 2765097
IUPAC Name 2-(prop-2-en-1-yloxy)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES C=CCON1C(=O)C2=CC=CC=C2C1=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 203.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Allyloxyphthalimide