missing translation for 'onlineSavingsMsg'
Learn More

N-(Allyloxycarbonyloxy)succinimide 97.0+%, TCI America™

Catalog No. p-7117828
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23025G 5 g
A230225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23025G Supplier TCI America Supplier No. A23025G
Only null left

Chemical Identifiers

CAS 135544-68-2
Molecular Formula C8H9NO5
Molecular Weight (g/mol) 199.162
InChI Key OIXALTPBNZNFLJ-UHFFFAOYSA-N
Synonym Allyl N-Succinimidyl Carbonate
PubChem CID 11788855
IUPAC Name (2,5-dioxopyrrolidin-1-yl) prop-2-enyl carbonate
SMILES C=CCOC(=O)ON1C(=O)CCC1=O

Specifications

Color White
Quantity 5 g
Formula Weight 199.16
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material N-(Allyloxycarbonyloxy)succinimide
TSCA No
Recommended Storage Refrigerator