missing translation for 'onlineSavingsMsg'
Learn More

N-Allyl-N-tert-butylamine 98.0+%, TCI America™

Catalog No. A251425G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A251425G 25 g
A2514100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A251425G Supplier TCI America Supplier No. A251425G

Chemical Identifiers

CAS 16486-68-3
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.204
InChI Key MDFOQCKFSUMLET-UHFFFAOYSA-N
Synonym N-tert-Butylallylamine
PubChem CID 566539
IUPAC Name 2-methyl-N-prop-2-enylpropan-2-amine
SMILES CC(C)(C)NCC=C

Specifications

Color Yellow
Boiling Point 112°C
Quantity 25 g
UN Number 2733
Formula Weight 113.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Allyl-N-tert-butylamine