accessibility menu, dialog, popup

UserName

CH3CH3H3CNHH2C

CAS RN 16486-68-3

IUPAC Name: tert-butyl(prop-2-en-1-yl)amine
Synonyms: tert-butyl(prop-2-en-1-yl)amine
Molecular Weight (g/mol): 113.2
Molecular Formula: C7H15N
InChi Key: MDFOQCKFSUMLET-UHFFFAOYSA-N
SMILES: CC(C)(C)NCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results