missing translation for 'onlineSavingsMsg'
Learn More

N-Acryloxysuccinimide, 99%

Catalog No. AC400300010
Change view
Click to view available options
Quantity:
1 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC400300010 1 g Glass bottle
AC400300250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC400300010 Supplier Thermo Scientific Chemicals Supplier No. 400300010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 38862-24-7 | C7H7NO4 | 169.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 38862-24-7
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.14
MDL Number MFCD00078261
InChI Key YXMISKNUHHOXFT-UHFFFAOYSA-N
Synonym 2,5-dioxopyrrolidin-1-yl acrylate, n-acryloxysuccinimide, n-succinimidyl acrylate, acrylic acid n-hydroxysuccinimide ester, 1-acryloyloxy-2,5-pyrrolidinedione, n-acryloyloxysuccinimide, 2,5-pyrrolidinedione, 1-1-oxo-2-propenyl oxy, 2,5-dioxopyrrolidin-1-yl prop-2-enoate, poly n-acryloxysuccinimide
PubChem CID 181508
IUPAC Name (2,5-dioxopyrrolidin-1-yl) prop-2-enoate
SMILES C=CC(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 69.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in diethyl ether,hexane,ethyl acetate
Formula Weight 169.14
Percent Purity 99%
Physical Form Crystals and/or Chunks
Chemical Name or Material N-Acryloxysuccinimide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only