missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyltryptamine 98.0+%, TCI America™

Catalog No. p-7118021
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A268525MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A268525MG Supplier TCI America Supplier No. A268525MG
Only null left

Chemical Identifiers

CAS 1016-47-3
Molecular Formula C12H14N2O
Molecular Weight (g/mol) 202.257
MDL Number MFCD00209910
InChI Key NVUGEQAEQJTCIX-UHFFFAOYSA-N
Synonym n-acetyltryptamine, n-2-1h-indol-3-yl ethyl acetamide, 3-2-n-acetylaminoethyl indole, acetamide, n-2-1h-indol-3-yl ethyl, chembl33171, n-2-1h-indol-3-yl-ethyl-acetamide, acetotryptamide, acetamide, n-2-indol-3-ylethyl
PubChem CID 70547
ChEBI CHEBI:55515
IUPAC Name N-[2-(1H-indol-3-yl)ethyl]acetamide
SMILES CC(=O)NCCC1=CNC2=CC=CC=C21

Specifications

Melting Point 76°C
Boiling Point 200°C
Quantity 25 mg
Formula Weight 202.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyltryptamine