accessibility menu, dialog, popup

UserName

CAS RN 1016-47-3

IUPAC Name: N-[2-(1H-indol-3-yl)ethyl]acetamide
Synonyms: N-acetyltryptamine
Molecular Weight (g/mol): 202.26
Molecular Formula: C12H14N2O
InChi Key: NVUGEQAEQJTCIX-UHFFFAOYSA-N
SMILES: CC(=O)NCCC1=CNC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results