missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylphthalimide 98.0+%, TCI America™

Catalog No. A010825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A010825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A010825G Supplier TCI America Supplier No. A010825G
Only null left

Chemical Identifiers

CAS 1971-49-9
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00023054
InChI Key INZUQGFQRYAKQQ-UHFFFAOYSA-N
Synonym n-acetylphthalimide, 2-acetylisoindoline-1,3-dione, acetylphthalimide, phthalimide, n-acetyl, 2-acetyl-1,3-dioxobenzo c azoline, n-acetylphthali-mide, 1h-isoindole-1,3 2h-dione, 2-acetyl, maybridge1_007322, acmc-209f0t
PubChem CID 243267
IUPAC Name 2-acetylisoindole-1,3-dione
SMILES CC(=O)N1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 137°C
Color White-Yellow
Quantity 25 g
Formula Weight 189.17
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetylphthalimide