accessibility menu, dialog, popup

UserName

H3CONOO

CAS RN 1971-49-9

IUPAC Name: 2-acetyl-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-acetylisoindole-1,3-dione
Molecular Weight (g/mol): 189.17
Molecular Formula: C10H7NO3
InChi Key: INZUQGFQRYAKQQ-UHFFFAOYSA-N
SMILES: CC(=O)N1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results