missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylneuraminic Acid Methyl Ester 95.0+%, TCI America™

Catalog No. A18211G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A18211G Supplier TCI America Supplier No. A18211G
Only null left

Chemical Identifiers

CAS 22900-11-4
Molecular Formula C12H21NO9
Molecular Weight (g/mol) 323.30
MDL Number MFCD08276447
InChI Key BKZQMWNJESHHSA-UHFFFAOYNA-N
Synonym Methyl 5-Acetamido-3,5-dideoxy-D-glycero-D-galacto-2-nonulopyranosonate
PubChem CID 7173831
IUPAC Name methyl 5-acetamido-2,4-dihydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylate
SMILES COC(=O)C1(O)CC(O)C(NC(C)=O)C(O1)C(O)C(O)CO

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 323.30
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetylneuraminic Acid Methyl Ester
TSCA No
Recommended Storage Freezer