missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylneuraminic Acid Hydrate 98.0+%, TCI America™

Supplier:  TCI America A06395G

Quantity 5 g

 View more versions of this product

Catalog No. A06395G


Only null left

Description

Description

Specifications

Chemical Identifiers

131-48-6
309.271
SQVRNKJHWKZAKO-LFIUDZTESA-N
126963458
CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)O)O
C11H19NO9
MFCD00006620
n-acetylneuramic acid, spectrum_000450, spectrum2_000559, spectrum4_000432, spectrum5_001326, spectrum2300147, 2s,4s,5s,6s-5-acetamido-2,4-dihydroxy-6-2r-1,2,3-trihydroxypropyl oxane-2-carboxylic acid
(4S,5S,6R)-5-acetamido-2,4-dihydroxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid

Specifications

N-Acetylneuraminic Acid Hydrate
186°C
C11H19NO9
MFCD00006620
SQVRNKJHWKZAKO-LFIUDZTESA-N
(4S,5S,6R)-5-acetamido-2,4-dihydroxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid
126963458
≥98.0% (T)
131-48-6
White-Yellow
5 g
n-acetylneuramic acid, spectrum_000450, spectrum2_000559, spectrum4_000432, spectrum5_001326, spectrum2300147, 2s,4s,5s,6s-5-acetamido-2,4-dihydroxy-6-2r-1,2,3-trihydroxypropyl oxane-2-carboxylic acid
CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)O)O
309.271
309.27
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6979

TSCA : Yes

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions