accessibility menu, dialog, popup

UserName

CAS RN 131-48-6

IUPAC Name: 5-acetamido-2,4-dihydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
Synonyms: acetylneuraminic acid
Molecular Weight (g/mol): 309.27
Molecular Formula: C11H19NO9
InChi Key: SQVRNKJHWKZAKO-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(O)CC(O)(OC1C(O)C(O)CO)C(O)=O
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)