missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylneuraminic Acid Hydrate 98.0+%, TCI America™

Catalog No. A0639100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0639100MG 100 mg
A06391G 1 g
A06395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0639100MG Supplier TCI America Supplier No. A0639100MG

Chemical Identifiers

CAS 131-48-6
Molecular Formula C11H19NO9
Molecular Weight (g/mol) 309.271
MDL Number MFCD00006620
InChI Key SQVRNKJHWKZAKO-LFIUDZTESA-N
Synonym n-acetylneuramic acid, spectrum_000450, spectrum2_000559, spectrum4_000432, spectrum5_001326, spectrum2300147, 2s,4s,5s,6s-5-acetamido-2,4-dihydroxy-6-2r-1,2,3-trihydroxypropyl oxane-2-carboxylic acid
PubChem CID 126963458
IUPAC Name (4S,5S,6R)-5-acetamido-2,4-dihydroxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid
SMILES CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)O)O

Specifications

Melting Point 186°C
Color White-Yellow
Quantity 100 mg
Formula Weight 309.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetylneuraminic Acid Hydrate
EINECSNumber (5)-6979
TSCA Yes
Recommended Storage Freezer