missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylglucosamine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015431
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015431 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015431 Supplier Merck Emd Millipore Supplier No. PHR14321G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

N-Acetylglucosamine (GlcNAc) is a monosaccharide, derived from glucose and undergoes linear polymerization by (1,4)-β-linkages to form the polymer chitin. It is also a component of heterogenous polysaccharides, such as murein and hyaluronic acid. N-acetylglucosamine exhibits anti-inflammatory properties and is considered as a potential drug in the treatment of osteoarthritis. Its antiarthritic mode of mechanism involves the suppression of IL-1β (Interleuken-1β)-induced NO (nitric oxide), cyclooxygenase-2 (COX-2), and IL-6 production by inhibition of expression of iNOS (inducible nitric oxide synthase) protein and mRNA.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 7512-17-6
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C8H15 NO6
MDL Number MFCD00061615
Melting Point 211°C
Synonym N-Acetyl-D-glucosamine; 2-Acetamido-2-deoxy-D-glucose; D-GlcNAc
Type Certified Reference Material
Quantity 1 g
Formula Weight 221.21
Grade Certified Reference Material