missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylcaprolactam, 99%

Catalog No. AAA1825714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1825714 25 g
AAA1825722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1825714 Supplier Thermo Scientific Chemicals Supplier No. A1825714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1888-91-1 | C8H13NO2 | 155.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1888-91-1
Molecular Formula C8H13NO2
Molecular Weight (g/mol) 155.20
MDL Number MFCD00003262
InChI Key QISSLHPKTCLLDL-UHFFFAOYSA-N
Synonym n-acetylcaprolactam, acetylcaprolactam, n-acetyl-6-caprolactam, 2h-azepin-2-one, 1-acetylhexahydro, acetylkaprolaktam, n-acetylhexanelactam, acetylkaprolaktam czech, 1-acetylhexahydro-2h-azepin-2-one, n-acetyl-epsilon-caprolactam, n-acetyl-.epsilon.-caprolactam
PubChem CID 15904
IUPAC Name 1-acetylazepan-2-one
SMILES CC(=O)N1CCCCCC1=O

Specifications

Density 1.094
Boiling Point 124°C (12 mmHg)
Flash Point 122°C (252°F)
Refractive Index 1.489
Quantity 25 g
Beilstein 119598
Formula Weight 155.2
Percent Purity 99%
Chemical Name or Material N-Acetylcaprolactam
Hazard Category H302-H319-H361
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P264b-P270-P280i-P281-P301+P312-P305+P351+P338-P308+P313-P330-P501c
EINECSNumber 217-565-6
RTECSNumber CM3599500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only