missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-O-tert-butyl-L-serine, 95%, Thermo Scientific Chemicals

Catalog No. p-7056365
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6523903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6523903 Supplier Thermo Scientific Chemicals Supplier No. H6523903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 77285-09-7
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.24
MDL Number MFCD00236770
InChI Key COMGVZVKADFEPL-UHFFFAOYNA-N
Synonym ac-ser tbu-oh, l-serine,n-acetyl-o-1,1-dimethylethyl, ambotzaaa1904, n-acetyl-o-t-butyl-l-serine, 2s-3-tert-butoxy-2-acetamidopropanoic acid
PubChem CID 7019612
IUPAC Name 3-(tert-butoxy)-2-acetamidopropanoic acid
SMILES CC(=O)NC(COC(C)(C)C)C(O)=O

Specifications

Color White
Beilstein 9047283
Quantity 1 g
Formula Weight 203.24
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-O-tert-butyl-L-serine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only