accessibility menu, dialog, popup

UserName

CAS RN 77285-09-7

IUPAC Name: 3-(tert-butoxy)-2-acetamidopropanoic acid
Synonyms: 3-(tert-butoxy)-2-acetamidopropanoic acid
Molecular Weight (g/mol): 203.24
Molecular Formula: C9H17NO4
InChi Key: COMGVZVKADFEPL-UHFFFAOYNA-N
SMILES: CC(=O)NC(COC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)