missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-tyrosine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014521
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014521 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014521 Supplier Merck Emd Millipore Supplier No. PHR11731G
N-Acetyl-L-tyrosine is an acetyl derivative of the amino acid, L-tyrosine. It is freely soluble in water compared to L-tyrosine and is commonly used as a parenteral nutrition supplement.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 537-55-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula 4-(HO)C6H4 CH2 CH(NHCOCH3)CO2 H
MDL Number MFCD00037190
Melting Point 149°C to 152°C
Type Certified Reference Material
Quantity 1 g
Formula Weight 223.23
Grade Certified Reference Material