Learn More
Thermo Scientific Chemicals N-Acetyl-L-tyrosine, 99%
CAS: 537-55-3 | C11H13NO4 | 223.23 g/mol
Supplier: Thermo Scientific Chemicals A1730718
Description
N-Acetyl-L-tyrosine is involved in catecholamine production. It can be used as a cell culture media component in the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Acetyl-L-tyrosine is involved in catecholamine production. It can be used as a cell culture media component in the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.
Solubility
Soluble in water (25 mg/ml), and ethanol.
Notes
Store at room temperature. Keep away from strong oxidizing agents.
Specifications
| 150°C to 151°C | |
| 537-55-3 | |
| MFCD00037190 | |
| n-acetyl-l-tyrosine, ac-tyr-oh, acetyl-l-tyrosine, n-acetyl-tyrosine, n-acetyltyrosine, acetyl tyrosine, l-n-acetyltyrosine, tyrosine, n-acetyl, l-tyrosine, n-acetyl, unii-da8g610zo5 | |
| CAHKINHBCWCHCF-UEQNJFAPNA-N | |
| (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoic acid | |
| 223.23 | |
| CHEBI:21563 | |
| 223.23 |
| N-Acetyl-L-tyrosine | |
| C11H13NO4 | |
| 2697172 | |
| Soluble in water (25mg/ml),and ethanol. | |
| CC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O | |
| 50 g | |
| 68310 | |
| +58° (c=5 in Ethanol) | |
| 99% |
Chemical Identifiers
| 537-55-3 | |
| 223.23 | |
| CAHKINHBCWCHCF-UEQNJFAPNA-N | |
| 68310 | |
| (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoic acid |
| C11H13NO4 | |
| MFCD00037190 | |
| n-acetyl-l-tyrosine, ac-tyr-oh, acetyl-l-tyrosine, n-acetyl-tyrosine, n-acetyltyrosine, acetyl tyrosine, l-n-acetyltyrosine, tyrosine, n-acetyl, l-tyrosine, n-acetyl, unii-da8g610zo5 | |
| CHEBI:21563 | |
| CC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O |
Safety and Handling
P280-P305+P351+P338-P310
H318
EINECSNumber : 208-671-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only