missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-L-tyrosine, 99%

Catalog No. p-7023592
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1730709 10 g
AAA1730718 50 g
AAA1730730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1730709 Supplier Thermo Scientific Chemicals Supplier No. A1730709

CAS: 537-55-3 | C11H13NO4 | 223.23 g/mol

N-Acetyl-L-tyrosine is involved in catecholamine production. It can be used as a cell culture media component in the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-L-tyrosine is involved in catecholamine production. It can be used as a cell culture media component in the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

Solubility
Soluble in water (25 mg/ml), and ethanol.

Notes
Store at room temperature. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 537-55-3
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00037190
InChI Key CAHKINHBCWCHCF-UEQNJFAPNA-N
Synonym n-acetyl-l-tyrosine, ac-tyr-oh, acetyl-l-tyrosine, n-acetyl-tyrosine, n-acetyltyrosine, acetyl tyrosine, l-n-acetyltyrosine, tyrosine, n-acetyl, l-tyrosine, n-acetyl, unii-da8g610zo5
PubChem CID 68310
ChEBI CHEBI:21563
IUPAC Name (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoic acid
SMILES CC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Melting Point 150°C to 151°C
Beilstein 2697172
Solubility Information Soluble in water (25mg/ml),and ethanol.
Quantity 10 g
Formula Weight 223.23
Optical Rotation +58° (c=5 in Ethanol)
Percent Purity 99%
Chemical Name or Material N-Acetyl-L-tyrosine
Hazard Category H318
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 208-671-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only