missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-L-tryptophan 98.0+%, TCI America™
Supplier: TCI America A012125G
Specifications
| N-Acetyl-L-tryptophan | |
| 186°C | |
| C13H14N2O3 | |
| 25 g | |
| DZTHIGRZJZPRDV-LBPRGKRZSA-N | |
| (2S)-2-acetamido-3-(1H-indol-3-yl)propanoic acid | |
| 700653 | |
| 246.27 | |
| Crystalline Powder |
| 1218-34-4 | |
| Red-White | |
| MFCD00065976 | |
| n-acetyl-l-tryptophan, ac-trp-oh, acetyl-l-tryptophan, acetyltryptophan, acetyl-l-trp, ac-try, s-n-acetyltryptophan, unii-u9264t8oae, tryptophan, n-acetyl, s-2-acetamido-3-1h-indol-3-yl propanoic acid | |
| CC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O | |
| 246.27 | |
| CHEBI:74640 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 1218-34-4 | |
| 246.27 | |
| DZTHIGRZJZPRDV-LBPRGKRZSA-N | |
| 700653 | |
| (2S)-2-acetamido-3-(1H-indol-3-yl)propanoic acid |
| C13H14N2O3 | |
| MFCD00065976 | |
| n-acetyl-l-tryptophan, ac-trp-oh, acetyl-l-tryptophan, acetyltryptophan, acetyl-l-trp, ac-try, s-n-acetyltryptophan, unii-u9264t8oae, tryptophan, n-acetyl, s-2-acetamido-3-1h-indol-3-yl propanoic acid | |
| CHEBI:74640 | |
| CC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O |
Safety and Handling
EINECSNumber : (9)-1601
RTECSNumber : YN6160000
TSCA : Yes