missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-tryptophan 98.0+%, TCI America™

Catalog No. p-7116729
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A01211G 1 g
A012125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A01211G Supplier TCI America Supplier No. A01211G
Only null left

Chemical Identifiers

CAS 1218-34-4
Molecular Formula C13H14N2O3
Molecular Weight (g/mol) 246.27
MDL Number MFCD00065976
InChI Key DZTHIGRZJZPRDV-LBPRGKRZSA-N
Synonym n-acetyl-l-tryptophan, ac-trp-oh, acetyl-l-tryptophan, acetyltryptophan, acetyl-l-trp, ac-try, s-n-acetyltryptophan, unii-u9264t8oae, tryptophan, n-acetyl, s-2-acetamido-3-1h-indol-3-yl propanoic acid
PubChem CID 700653
ChEBI CHEBI:74640
IUPAC Name (2S)-2-acetamido-3-(1H-indol-3-yl)propanoic acid
SMILES CC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Melting Point 186°C
Color Red-White
Quantity 1 g
Formula Weight 246.27
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-L-tryptophan
EINECSNumber (9)-1601
RTECSNumber YN6160000
TSCA Yes