missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-cysteine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014904
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014904 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014904 Supplier Merck Emd Millipore Supplier No. PHR10981G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical (small molecule)

Specifications

CAS 616-91-1
Linear Formula HSCH2 CH(NHCOCH3)CO2 H
MDL Number MFCD00004880
Melting Point 106°C to 108°C
Synonym LNAC; NAC
Type Certified Reference Material
Quantity 1 g
Formula Weight 163.19
Grade Certified Reference Material