missing translation for 'onlineSavingsMsg'
Learn More

N-acetyl-L-cysteine, >96%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210009880

 View more versions of this product

Catalog No. ICN10009880

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-acetyl-L-cysteine
White
C5H9NO3S
n-acetyl-l-cysteine, acetylcysteine, n-acetylcysteine, mercapturic acid, acetadote, l-acetylcysteine, broncholysin, fluimucil, mucomyst, fluprowit
CC(=O)NC(CS)C(=O)O
(2R)-2-acetamido-3-sulfanylpropanoic acid
163.191
CHEBI:28939
Crystalline Powder
616-91-1
>96%
MFCD00004880
PWKSKIMOESPYIA-BYPYZUCNSA-N
1.295g/cm3 (L)
100 g
12035
163.2

Chemical Identifiers

616-91-1
163.191
PWKSKIMOESPYIA-BYPYZUCNSA-N
12035
(2R)-2-acetamido-3-sulfanylpropanoic acid
C5H9NO3S
MFCD00004880
n-acetyl-l-cysteine, acetylcysteine, n-acetylcysteine, mercapturic acid, acetadote, l-acetylcysteine, broncholysin, fluimucil, mucomyst, fluprowit
CHEBI:28939
CC(=O)NC(CS)C(=O)O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions