missing translation for 'onlineSavingsMsg'
Learn More

N-acetyl-L-cysteine, >96%, MP Biomedicals™

Catalog No. p-4403619
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10009825 25 g
ICN10009850 50 g
ICN10009880 100 g
ICN10009890 500 g
ICN10009891 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN10009825 Supplier MP Biomedicals Inc Supplier No. 0210009825
Only null left

Chemical Identifiers

CAS 616-91-1
Molecular Formula C5H9NO3S
Molecular Weight (g/mol) 163.191
MDL Number MFCD00004880
InChI Key PWKSKIMOESPYIA-BYPYZUCNSA-N
Synonym n-acetyl-l-cysteine, acetylcysteine, n-acetylcysteine, mercapturic acid, acetadote, l-acetylcysteine, broncholysin, fluimucil, mucomyst, fluprowit
PubChem CID 12035
ChEBI CHEBI:28939
IUPAC Name (2R)-2-acetamido-3-sulfanylpropanoic acid
SMILES CC(=O)NC(CS)C(=O)O

Specifications

Color White
Assay Percent Range >96%
Density 1.295g/cm3 (L)
Quantity 25 g
Formula Weight 163.2
Physical Form Crystalline Powder
Chemical Name or Material N-acetyl-L-cysteine