missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-acetyl-L-arginine dihydrate, >99%, MP Biomedicals™
Manufacturer: Mp Biomedicals 10007810
Specifications
N-acetyl-L-arginine dihydrate | |
>99% | |
MFCD00019733 | |
SNEIUMQYRCDYCH-LURJTMIESA-N | |
(2S)-2-acetamido-5-(diaminomethylideneamino)pentanoic acid | |
67427 | |
252.27 | |
White |
155-84-0 | |
C8H16N4O3 | |
ac-arg-oh | |
CC(=O)NC(CCCN=C(N)N)C(=O)O | |
216.241 | |
CHEBI:40521 | |
Powder | |
10g |
Chemical Identifiers
155-84-0 | |
216.241 | |
SNEIUMQYRCDYCH-LURJTMIESA-N | |
67427 | |
(2S)-2-acetamido-5-(diaminomethylideneamino)pentanoic acid |
C8H16N4O3 | |
MFCD00019733 | |
ac-arg-oh | |
CHEBI:40521 | |
CC(=O)NC(CCCN=C(N)N)C(=O)O |
Safety and Handling
EINECSNumber : 205-846-6