CAS RN 155-84-0
CAS RN 155-84-0
IUPAC Name:
(2S)-5-[(diaminomethylidene)amino]-2-acetamidopentanoic acid
Synonyms:
(2S)-5-[(diaminomethylidene)amino]-2-acetamidopentanoic acid
Molecular Weight (g/mol):
216.24
Molecular Formula:
C8H16N4O3
InChi Key:
SNEIUMQYRCDYCH-LURJTMIESA-N
SMILES:
CC(=O)N[C@@H](CCCN=C(N)N)C(O)=O
1
–
1
of
1
results
1
N-acetyl-L-arginine dihydrate, >99%, MP Biomedicals™
CAS: 155-84-0 Molecular Formula: C8H16N4O3 Molecular Weight (g/mol): 216.241 MDL Number: MFCD00019733 InChI Key: SNEIUMQYRCDYCH-LURJTMIESA-N Synonym: n-acetyl-l-arginine,n-alpha-l-acetyl-arginine,n2-acetyl-l-arginine,arginine, n2-acetyl,ac-arg-oh.2h2o,unii-tq7dl04cae,s-2-acetamido-5-guanidinopentanoic acid,ac-arg-oh,n-a-acetyl-l-arginine,nalpha-acetyl-l-arginine PubChem CID: 67427 ChEBI: CHEBI:40521 IUPAC Name: (2S)-2-acetamido-5-(diaminomethylideneamino)pentanoic acid SMILES: CC(=O)NC(CCCN=C(N)N)C(=O)O