missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-epsilon-caprolactam 98.0+%, TCI America™

Catalog No. A0081500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0081500G 500 g
A008125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0081500G Supplier TCI America Supplier No. A0081500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1888-91-1
Molecular Formula C8H13NO2
Molecular Weight (g/mol) 155.20
MDL Number MFCD00003262
InChI Key QISSLHPKTCLLDL-UHFFFAOYSA-N
Synonym n-acetylcaprolactam, acetylcaprolactam, n-acetyl-6-caprolactam, 2h-azepin-2-one, 1-acetylhexahydro, acetylkaprolaktam, n-acetylhexanelactam, acetylkaprolaktam czech, 1-acetylhexahydro-2h-azepin-2-one, n-acetyl-epsilon-caprolactam, n-acetyl-.epsilon.-caprolactam
PubChem CID 15904
IUPAC Name 1-acetylazepan-2-one
SMILES CC(=O)N1CCCCCC1=O

Specifications

Color Yellow
Boiling Point 134°C
Quantity 500 g
Formula Weight 155.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Acetyl-epsilon-caprolactam
EINECSNumber (5)-3875
RTECSNumber CM3599500
TSCA Yes