missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-DL-tryptophan 98.0+%, TCI America™
$38.51 - $265.67
Chemical Identifiers
| CAS | 87-32-1 |
|---|---|
| Molecular Formula | C13H14N2O3 |
| Molecular Weight (g/mol) | 246.27 |
| MDL Number | MFCD00005644 |
| InChI Key | DZTHIGRZJZPRDV-UHFFFAOYSA-N |
| Synonym | n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl |
| PubChem CID | 2002 |
| ChEBI | CHEBI:70976 |
| IUPAC Name | 2-acetamido-3-(1H-indol-3-yl)propanoic acid |
| SMILES | CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O |
Chemical Identifiers
| 87-32-1 | |
| 246.27 | |
| DZTHIGRZJZPRDV-UHFFFAOYSA-N | |
| 2002 | |
| 2-acetamido-3-(1H-indol-3-yl)propanoic acid |
| C13H14N2O3 | |
| MFCD00005644 | |
| n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl | |
| CHEBI:70976 | |
| CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O |
Specifications
| 87-32-1 | |
| White | |
| MFCD00005644 | |
| n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl | |
| CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O | |
| 246.27 | |
| CHEBI:70976 | |
| ≥98.0% (HPLC,T) | |
| N-Acetyl-DL-tryptophan |
| 205°C | |
| C13H14N2O3 | |
| 1 g | |
| DZTHIGRZJZPRDV-UHFFFAOYSA-N | |
| 2-acetamido-3-(1H-indol-3-yl)propanoic acid | |
| 2002 | |
| 246.27 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-1601
TSCA : Yes