missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-DL-tryptophan, 99%

CAS: 87-32-1 | C13H14N2O3 | 246.27 g/mol

$141.22 - $418.38

Chemical Identifiers

CAS 87-32-1
Molecular Formula C13H14N2O3
Molecular Weight (g/mol) 246.27
MDL Number MFCD00005644
InChI Key DZTHIGRZJZPRDV-UHFFFAOYSA-N
Synonym n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl
PubChem CID 2002
ChEBI CHEBI:70976
IUPAC Name 2-acetamido-3-(1H-indol-3-yl)propanoic acid
SMILES CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1756214
View Documents
Thermo Scientific Chemicals
A1756214
25 g
Each for $141.22
Only null left
 
AAA1756222
View Documents
Thermo Scientific Chemicals
A1756222
100 g
Each for $418.38
Only null left
 
Description

Description

N-Acetyl-DL-tryptophan is produced via chemical synthesis using the standard amino acid, tryptophan. It can be used in downstream applications of biopharmaceutical production as a transfer agent or stabilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-DL-tryptophan is produced via chemical synthesis using the standard amino acid, tryptophan. It can be used in downstream applications of biopharmaceutical production as a transfer agent or stabilizer.

Solubility
Soluble in 1N Sodium Hydroxide, water, and methanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

87-32-1
246.27
DZTHIGRZJZPRDV-UHFFFAOYSA-N
2002
2-acetamido-3-(1H-indol-3-yl)propanoic acid
C13H14N2O3
MFCD00005644
n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl
CHEBI:70976
CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

∼205°C (decomposition)
C13H14N2O3
89478
Soluble in 1N Sodium Hydroxide,water,and methanol.
CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O
25 g
2002
246.27
N-Acetyl-DL-tryptophan
87-32-1
MFCD00005644
n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl
DZTHIGRZJZPRDV-UHFFFAOYSA-N
2-acetamido-3-(1H-indol-3-yl)propanoic acid
246.27
CHEBI:70976
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-739-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only