missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-DL-tryptophan, 99%

Catalog No. AAA1756214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1756214 25 g
AAA1756222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1756214 Supplier Thermo Scientific Chemicals Supplier No. A1756214
Only null left

CAS: 87-32-1 | C13H14N2O3 | 246.27 g/mol

N-Acetyl-DL-tryptophan is produced via chemical synthesis using the standard amino acid, tryptophan. It can be used in downstream applications of biopharmaceutical production as a transfer agent or stabilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-DL-tryptophan is produced via chemical synthesis using the standard amino acid, tryptophan. It can be used in downstream applications of biopharmaceutical production as a transfer agent or stabilizer.

Solubility
Soluble in 1N Sodium Hydroxide, water, and methanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 87-32-1
Molecular Formula C13H14N2O3
Molecular Weight (g/mol) 246.27
MDL Number MFCD00005644
InChI Key DZTHIGRZJZPRDV-UHFFFAOYSA-N
Synonym n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl
PubChem CID 2002
ChEBI CHEBI:70976
IUPAC Name 2-acetamido-3-(1H-indol-3-yl)propanoic acid
SMILES CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Melting Point ∼205°C (decomposition)
Beilstein 89478
Solubility Information Soluble in 1N Sodium Hydroxide,water,and methanol.
Quantity 25 g
Formula Weight 246.27
Percent Purity 99%
Chemical Name or Material N-Acetyl-DL-tryptophan
EINECSNumber 201-739-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only