missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-DL-2-aminobutyric Acid 97.0+%, TCI America™

Catalog No. p-7117090
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08535G 5 g
A085325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A08535G Supplier TCI America Supplier No. A08535G
Only null left

Chemical Identifiers

CAS 7211-57-6
Molecular Formula C6H11NO3
Molecular Weight (g/mol) 145.158
MDL Number MFCD00020430
InChI Key WZVZUKROCHDMDT-UHFFFAOYSA-N
Synonym 2-Acetamidobutyric Acid
PubChem CID 306107
IUPAC Name 2-acetamidobutanoic acid
SMILES CCC(C(=O)O)NC(=O)C

Specifications

Melting Point 132°C
Color White
Quantity 5 g
Formula Weight 145.16
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-DL-2-aminobutyric Acid