accessibility menu, dialog, popup

UserName

CAS RN 7211-57-6

H3COHOHNOCH3

CAS RN 7211-57-6

IUPAC Name: 2-acetamidobutanoic acid
Synonyms: 2-acetamidobutanoic acid
Molecular Weight (g/mol): 145.16
Molecular Formula: C6H11NO3
InChi Key: WZVZUKROCHDMDT-UHFFFAOYNA-N
SMILES: CCC(NC(C)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results