missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-DL-2-aminobutyric Acid 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 132°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 145.16 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-Acetyl-DL-2-aminobutyric Acid |
Safety and Handling
| TSCA | No |