missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-D-sphingosine, 98%

$224.59 - $355.15

Chemical Identifiers

CAS 3102-57-6
Molecular Formula C20H39NO3
Synonym Acetyl ceramide; C2-Ceramide
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ60534LB0
View Documents Promotion Details
Thermo Scientific Chemicals
J60534LB0
5 mg
Each for $224.59
Only null left
 
AAJ60534MA
View Documents Promotion Details
Thermo Scientific Chemicals
J60534MA
10 mg
Each for $355.15
Only null left
 
Description

Description

Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells. Activates protein phosphatases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells
  • Activates protein phosphatases
Specifications

Chemical Identifiers

3102-57-6
C20H39NO3
Acetyl ceramide; C2-Ceramide

Specifications

White
Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells. Activates protein phosphatases
C20H39NO3
Acetyl ceramide; C2-Ceramide
Soluble in ethanol, methanol, chloroform or DMSO at approximately 10mg/ml
Powder
CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](CO)NC(C)=O
341.54
-30°C to -10°C
3102-57-6
341.53g/mol
77°C to 78°C
Signal Transduction Reagent
BLTCBVOJNNKFKC-QUDYQQOWSA-N
N-[(2S,3R,4E)-1,3-dihydroxyoctadec-4-en-2-yl]acetamide
98%
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only