missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-sphingosine, 98%, Thermo Scientific Chemicals

Catalog No. AAJ60534MA
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60534MA 10 mg
AAJ60534LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60534MA Supplier Thermo Scientific Chemicals Supplier No. J60534MA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3102-57-6 | C20H39NO3, C20H39NO3 | 341.54 g/mol

Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells. Activates protein phosphatases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells
  • Activates protein phosphatases

Chemical Identifiers

CAS 3102-57-6
Molecular Formula C20H39NO3
Synonym Acetyl ceramide; C2-Ceramide

Specifications

Quantity 10 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells. Activates protein phosphatases
Formula Weight 341.53g/mol
Melting Point 77°C to 78°C
Solubility Soluble in ethanol, methanol, chloroform or DMSO at approximately 10mg/ml
Product Type Signal Transduction Reagent
Form Powder
Assay 98%

RUO – Research Use Only