missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-phenylalanine 98.0+%, TCI America™

Catalog No. p-7116719
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A01051G 1 g
A01055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A01051G Supplier TCI America Supplier No. A01051G
Only null left

Chemical Identifiers

CAS 10172-89-1
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.23
MDL Number MFCD00002664
InChI Key CBQJSKKFNMDLON-SNVBAGLBSA-N
Synonym n-acetyl-d-phenylalanine, acetyl-d-phenylalanine, ac-d-phe-oh, n-acetyl-3-phenyl-d-alanine, n-acetylphenylalanine, d, d-phenylalanine, n-acetyl, r-2-acetamido-3-phenylpropanoic acid, 2r-2-acetamido-3-phenylpropanoic acid, n-acetylphenylalanine #, n-ac-d-phe-oh
PubChem CID 101184
ChEBI CHEBI:28203
IUPAC Name (2R)-2-acetamido-3-phenylpropanoic acid
SMILES CC(=O)N[C@H](CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 167°C
Color White
Quantity 1 g
Formula Weight 207.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-D-phenylalanine
EINECSNumber (9)-1617
TSCA No