accessibility menu, dialog, popup

UserName

H3COHN(R)OHO

CAS RN 10172-89-1

IUPAC Name: (2R)-2-acetamido-3-phenylpropanoic acid
Synonyms: N-acetyl-D-phenylalanine
Molecular Weight (g/mol): 207.23
Molecular Formula: C11H13NO3
InChi Key: CBQJSKKFNMDLON-SNVBAGLBSA-N
SMILES: CC(=O)N[C@H](CC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results