missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-D-phenylalanine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 167°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 207.23 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-Acetyl-D-phenylalanine |
Safety and Handling
| EINECSNumber | (9)-1617 |
| TSCA | No |