missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-mannosamine Monohydrate 98.0+%, TCI America™

Catalog No. A21601G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
A21601G 1 g
A21605G 5 g
2 options

Catalog No. A21601G

Supplier: TCI America A21601G

Only null left

Chemical Identifiers

CAS 3615-17-6
Molecular Formula C8H15NO6
MDL Number MFCD00136044
Synonym 2-Acetamido-2-deoxy-D-mannopyranose

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 1 g
Formula Weight 221.21
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-D-mannosamine Monohydrate