accessibility menu, dialog, popup

UserName

CAS RN 3615-17-6

CH3ONH(S)OHO(R)OH(S)HO(R)HO

CAS RN 3615-17-6

IUPAC Name: N-[(3S,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Synonyms: ManNAc N-acetyl-D-mannosamine 2-acetamido-2-deoxy-D-mannose
Molecular Weight (g/mol): 221.21
Molecular Formula: C8H15NO6
InChi Key: OVRNDRQMDRJTHS-ZTVVOAFPSA-N
SMILES: CC(=O)N[C@@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results