missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-D-leucine, 99%
CAS: 19764-30-8 | C8H15NO3 | 173.21 g/mol
$508.41 - $1643.49
Chemical Identifiers
| CAS | 19764-30-8 |
|---|---|
| Molecular Formula | C8H15NO3 |
| Molecular Weight (g/mol) | 173.21 |
| MDL Number | MFCD00066069 |
| InChI Key | WXNXCEHXYPACJF-JLDDOWRYNA-N |
| Synonym | n-acetyl-d-leucine, ac-d-leu-oh, d-leucine, n-acetyl, n-acethy-d-leucine, n-acetyl-r-leucine, unii-91wu82ga22, 2,n-acetyl-d-leucine, r-2-acetamido-4-methylpentanoic acid, 2r-2-acetamido-4-methylpentanoic acid, acetylleucine, d |
| PubChem CID | 1241420 |
| IUPAC Name | (2R)-2-acetamido-4-methylpentanoic acid |
| SMILES | CC(C)C[C@@H](NC(C)=O)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2704606
|
Thermo Scientific Chemicals
H2704606 |
5 g |
Each for $508.41
|
|
|||||
|
AAH2704614
|
Thermo Scientific Chemicals
H2704614 |
25 g |
Each for $1,643.49
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 19764-30-8 | |
| 173.21 | |
| WXNXCEHXYPACJF-JLDDOWRYNA-N | |
| 1241420 | |
| CC(C)C[C@@H](NC(C)=O)C(O)=O |
| C8H15NO3 | |
| MFCD00066069 | |
| n-acetyl-d-leucine, ac-d-leu-oh, d-leucine, n-acetyl, n-acethy-d-leucine, n-acetyl-r-leucine, unii-91wu82ga22, 2,n-acetyl-d-leucine, r-2-acetamido-4-methylpentanoic acid, 2r-2-acetamido-4-methylpentanoic acid, acetylleucine, d | |
| (2R)-2-acetamido-4-methylpentanoic acid |
Specifications
| 19764-30-8 | |
| MFCD00066069 | |
| WXNXCEHXYPACJF-JLDDOWRYNA-N | |
| (2R)-2-acetamido-4-methylpentanoic acid | |
| 173.21 | |
| 173.21 | |
| N-Acetyl-D-leucine |
| C8H15NO3 | |
| n-acetyl-d-leucine, ac-d-leu-oh, d-leucine, n-acetyl, n-acethy-d-leucine, n-acetyl-r-leucine, unii-91wu82ga22, 2,n-acetyl-d-leucine, r-2-acetamido-4-methylpentanoic acid, 2r-2-acetamido-4-methylpentanoic acid, acetylleucine, d | |
| CC(C)C[C@@H](NC(C)=O)C(O)=O | |
| 5 g | |
| 1241420 | |
| 99% |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only