missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-D-glucosamine, 98+%

N-Acetyl-D-glucosamine, CAS # 72-87-7, is the D isomer of the glucose-derived monomer found in structural polysaccharides.

$58.08 - $254.21

Chemical Identifiers

CAS 7512-17-6
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.21
MDL Number MFCD00061615
InChI Key OVRNDRQMDRJTHS-WZWLWIOANA-N
IUPAC Name N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1304709
View Documents
Thermo Scientific Chemicals
A1304709
10 g N/A
Only null left
 
AAA1304718
View Documents
Thermo Scientific Chemicals
A1304718
50 g N/A
Only null left
 
AAA1304722
View Documents
Thermo Scientific Chemicals
A1304722
100 g N/A
Only null left
 
Description

Description

N-Acetyl-D-glucosamine is used in the identification and characterization of N-acetyl-beta-D-hexoaminidase. It is also used in the treatment of osteoarthritis and inflammatory bowel disease (IBD), including ulcerative colitis and Crohn's disease. Further, it is used as a substrate in sialic acid production, cosmetics and in drug development research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• N-Acetyl-D-glucosamine is the acetylated form of glucosamine, a glucose derivative

• It has a role as a prokaryotic metabolism and monomer used in chitin, hyaluronic acid, and polymers within the cell wall

Application

• N-Acetyl-D-Glucosamine can be used for molecular binding studies; research has shown that it attaches to certain microbes and their pathogens

• It is useful for biochemical characterization studies of related glucosamines

Specifications

Chemical Identifiers

7512-17-6
221.21
OVRNDRQMDRJTHS-WZWLWIOANA-N
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O
C8H15NO6
MFCD00061615
N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide

Specifications

7512-17-6
≥98%
MFCD00061615
1727589
Soluble in concentrated hydrochloric acid,sulfuric acid,phosphoric acid and formic acid. Insoluble in water,dilute acids,dilute alkalis,concentrated alkalis and organic solvents.
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O
221.21
N-Acetyl-D-glucosamine
213°C (decomposition)
C8H15NO6
10 g
Hygroscopic
OVRNDRQMDRJTHS-WZWLWIOANA-N
N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
221.21
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-368-2

TSCA : Yes

storageNote1 : Hygroscopic

Recommended Storage : Keep cold; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only