missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-D-glucosamine, 98+%

N-Acetyl-D-glucosamine, CAS # 72-87-7, is the D isomer of the glucose-derived monomer found in structural polysaccharides.

Supplier:  Thermo Scientific Chemicals A1304722

 View more versions of this product

Catalog No. AAA1304722

Only null left
Explore available promotions

Description

Description

N-Acetyl-D-glucosamine is used in the identification and characterization of N-acetyl-beta-D-hexoaminidase. It is also used in the treatment of osteoarthritis and inflammatory bowel disease (IBD), including ulcerative colitis and Crohn's disease. Further, it is used as a substrate in sialic acid production, cosmetics and in drug development research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• N-Acetyl-D-glucosamine is the acetylated form of glucosamine, a glucose derivative

• It has a role as a prokaryotic metabolism and monomer used in chitin, hyaluronic acid, and polymers within the cell wall

Application

• N-Acetyl-D-Glucosamine can be used for molecular binding studies; research has shown that it attaches to certain microbes and their pathogens

• It is useful for biochemical characterization studies of related glucosamines

Specifications

Specifications

N-Acetyl-D-glucosamine
213°C (decomposition)
C8H15NO6
100 g
Hygroscopic
OVRNDRQMDRJTHS-WZWLWIOANA-N
N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
221.21
Powder
7512-17-6
Glass bottle
MFCD00061615
1727589
Soluble in concentrated hydrochloric acid,sulfuric acid,phosphoric acid and formic acid. Insoluble in water,dilute acids,dilute alkalis,concentrated alkalis and organic solvents.
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O
221.21
≥98%

Chemical Identifiers

7512-17-6
221.21
OVRNDRQMDRJTHS-WZWLWIOANA-N
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O
C8H15NO6
MFCD00061615
N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-368-2

TSCA : Yes

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only